Assay Details
Target Gene Details
Entrez Gene ID: | 57794 |
Gene Name: | SURP and G-patch domain containing 1 |
Gene Aliases: |
F23858, RBP, SF4 |
Location: |
Chr.19:19276513-19320512 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGP1 | NM_172231.3 | 10 | 1470 | NP_757386.2 |
| XM_005260002.2 | 9 | 1485 | XP_005260059.1 | |
| XM_011528155.2 | 9 | 1363 | XP_011526457.1 | |
| AF521128.1 | 10 | 1470 | AAN77123.1 | |
| AK092745.1 | 2 | 1412 | ||
| AK096152.1 | 9 | 1559 | ||
| AK292085.1 | 10 | 1467 | ||
| AL137286.1 | 4 | 758 | CAB70678.1 | |
| AL713757.1 | 4 | 758 | CAD28528.1 | |
| AY072916.1 | 10 | 1646 | AAL68960.1 | |
| AY072917.1 | 10 | 1450 | AAL68961.1 | |
| BC063784.1 | 10 | 1448 | AAH63784.1 | |
| BC142988.1 | 10 | 1467 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map