Assay Details
Target Gene Details
Entrez Gene ID: | 54938 |
Gene Name: | seryl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
SARS, SARSM, SERS, SYS, SerRS, SerRSmt, mtSerRS |
Location: |
Chr.19:38915264-38930896 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SARS2 | NM_001145901.1 | NP_001139373.1 | ||
| NM_017827.3 | NP_060297.1 | |||
| AB029948.1 | BAA99557.1 | |||
| AK000457.1 | BAA91176.1 | |||
| AK098522.1 | ||||
| AK293414.1 | ||||
| AK296152.1 | ||||
| BC042912.1 | AAH42912.1 |
More Information
Set Membership: |
|

Genomic Map