Assay Details
Target Gene Details
Entrez Gene ID: | 157567 |
Gene Name: | ankyrin repeat domain 46 |
Gene Aliases: |
ANK-S, GENX-115279 |
Location: |
Chr.8:100509752-100559786 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD46 | NM_001270377.1 | 3 | 409 | NP_001257306.1 |
| NM_001270378.1 | 3 | 1007 | NP_001257307.1 | |
| NM_001270379.1 | 3 | 409 | NP_001257308.1 | |
| NM_198401.3 | 4 | 514 | NP_940683.1 | |
| XM_006716509.1 | 3 | 550 | XP_006716572.1 | |
| XM_017013106.1 | 3 | 472 | XP_016868595.1 | |
| AB077205.1 | 4 | 512 | BAD23947.1 | |
| AK095235.1 | 3 | 384 | ||
| AL832842.1 | 3 | 369 | CAI46169.1 | |
| BC050396.1 | 3 | 365 | AAH50396.1 | |
| BC060843.1 | 3 | 382 | AAH60843.1 | |
| DA503253.1 | 3 | 409 |

Genomic Map