Assay Details
Target Gene Details
Entrez Gene ID: | 80204 |
Gene Name: | F-box protein 11 |
Gene Aliases: |
FBX11, PRMT9, UBR6, UG063H01, VIT1 |
Location: |
Chr.2:47806920-47905793 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO11 | NM_001190274.1 | 23 | 3424 | NP_001177203.1 |
| NM_025133.4 | 23 | 3213 | NP_079409.3 | |
| XM_005264572.4 | 22 | 3499 | XP_005264629.1 | |
| XM_005264573.4 | 22 | 3452 | XP_005264630.1 | |
| XM_017005015.1 | 23 | 3371 | XP_016860504.1 | |
| XM_017005016.1 | 22 | 3318 | XP_016860505.1 | |
| XM_017005017.1 | 23 | 3237 | XP_016860506.1 | |
| AF264714.1 | 4 | 2440 | AAF76888.1 | |
| AK025477.1 | 8 | 1416 | BAB15143.1 | |
| AK097734.1 | 12 | 1961 | ||
| AL117620.1 | 6 | 1195 | CAB56019.1 | |
| BC012728.2 | 9 | 1497 | AAH12728.2 | |
| BC043258.1 | 23 | 3315 | AAH43258.2 | |
| BC130445.1 | 23 | 3149 | ||
| BC136480.1 | 23 | 3149 |
More Information
Set Membership: |
|

Genomic Map