Assay Details
Target Gene Details
Entrez Gene ID: | 25974 |
Gene Name: | methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria |
Gene Aliases: |
cblC |
Location: |
Chr.1:45500184-45511266 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMACHC | NM_015506.2 | 4 | 2648 | NP_056321.2 |
| XM_005270724.4 | 3 | 2407 | XP_005270781.1 | |
| XM_011541204.2 | 4 | 2557 | XP_011539506.1 | |
| AK098537.1 | ||||
| AL080062.1 | 5 | 2042 | CAB45693.2 | |
| BU632364.1 | 1 | 219 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
| nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map