Assay Details
Target Gene Details
Entrez Gene ID: | 5704 |
Gene Name: | proteasome 26S subunit, ATPase 4 |
Gene Aliases: |
MIP224, RPT3, S6, TBP-7, TBP7 |
Location: |
Chr.19:39971005-39981764 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC4 | NM_006503.3 | NP_006494.1 | ||
| NM_153001.2 | NP_694546.1 | |||
| AF020736.1 | AAC32612.1 | |||
| AF038965.1 | AAC26843.1 | |||
| AK290285.1 | ||||
| AK310135.1 | 9 | 1121 | ||
| AK313183.1 | ||||
| BC000343.2 | AAH00343.1 | |||
| BC010396.1 | AAH10396.1 | |||
| BC014488.1 | AAH14488.1 | |||
| BT007232.1 | AAP35896.1 | |||
| U27515.1 | AAC99817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758760 | Chr.19:39737264 - 40015223 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map