Assay Details
Target Gene Details
Entrez Gene ID: | 83642 |
Gene Name: | selenoprotein O |
Gene Aliases: |
- |
Location: |
Chr.22:50200979-50217616 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SELO | NM_031454.1 | 7 | 1659 | NP_113642.1 |
| AY324823.1 | 7 | 1659 | AAP85540.1 | |
| BC001099.2 | 7 | 1149 | AAH01099.3 | |
| BC020510.1 | 6 | 970 | AAH20510.1 | |
| BC110866.1 | 7 | 1629 | AAI10867.1 | |
| CR456464.1 | 7 | 1175 | CAG30350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955199 | Chr.22:50149372 - 50320271 on Build GRCh38 | Deletion |
|
| nsv471223 | Chr.22:50190317 - 50271919 on Build GRCh38 | Loss |
|
| dgv781n67 | Chr.22:50153213 - 50314433 on Build GRCh38 | Gain |
|

Genomic Map