Assay Details
Target Gene Details
Entrez Gene ID: | 5270 |
Gene Name: | serpin family E member 2 |
Gene Aliases: |
GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI |
Location: |
Chr.2:223975048-224039319 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINE2 | NM_001136528.1 | 9 | 2069 | NP_001130000.1 |
| NM_001136530.1 | 9 | 1999 | NP_001130002.1 | |
| NM_006216.3 | 9 | 2072 | NP_006207.1 | |
| NR_073116.1 | 10 | 2487 | ||
| XM_005246641.2 | 9 | 2161 | XP_005246698.1 | |
| XM_017004329.1 | 9 | 2053 | XP_016859818.1 | |
| XM_017004330.1 | 9 | 1890 | XP_016859819.1 | |
| XM_017004331.1 | 9 | 2050 | XP_016859820.1 | |
| XM_017004332.1 | 9 | 1887 | XP_016859821.1 | |
| AI754402.1 | 1 | 125 | ||
| AJ293400.1 | ||||
| AK225823.1 | 9 | 2037 | ||
| BC015663.2 | 9 | 2029 | AAH15663.1 | |
| BC042628.1 | 10 | 2486 | AAH42628.1 | |
| BX360097.2 | 1 | 642 | ||
| BX647459.1 | 2 | 5904 | ||
| CR746616.1 | 1 | 207 | ||
| DB325994.1 | 1 | 188 |
More Information
Set Membership: |
|

Genomic Map