Assay Details
Target Gene Details
Entrez Gene ID: | 219833 |
Gene Name: | chromosome 11 open reading frame 45 |
Gene Aliases: |
- |
Location: |
Chr.11:128899565-128909520 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf45 | NM_001256088.1 | 4 | 1316 | NP_001243017.1 |
| NM_145013.2 | 4 | 1305 | NP_659450.1 | |
| NR_045767.1 | 4 | 1239 | ||
| XM_011542654.2 | 4 | 1235 | XP_011540956.1 | |
| XM_011542655.2 | 4 | 1197 | XP_011540957.1 | |
| AK125634.1 | 4 | 1213 | ||
| BC025756.1 | 4 | 1305 | AAH25756.1 | |
| BX956612.1 |
Target Gene Details
Entrez Gene ID: | 3762 |
Gene Name: | potassium voltage-gated channel subfamily J member 5 |
Gene Aliases: |
CIR, GIRK4, KATP1, KIR3.4, LQT13 |
Location: |
Chr.11:128891418-128921163 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNJ5 | NM_000890.3 | NP_000881.3 | ||
| AK312837.1 | ||||
| BC069386.1 | AAH69386.1 | |||
| BC069482.1 | AAH69482.1 | |||
| BC069499.1 | AAH69499.1 | |||
| BC069571.1 | AAH69571.1 | |||
| BC074838.2 | AAH74838.2 | |||
| BC074839.2 | AAH74839.2 | |||
| D50134.1 | BAA08814.1 | |||
| U39195.1 | AAB53093.1 |
More Information
Set Membership: |
|

Genomic Map