Assay Details
Target Gene Details
| Entrez Gene ID: | 219285 | 
| Gene Name: | sterile alpha motif domain containing 9 like | 
| Gene Aliases: | C7orf6, DRIF2, UEF1 | 
| Location: | Chr.7:93130054-93148401 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| SAMD9L | NM_001303496.1 | 5 | 6017 | NP_001290425.1 | 
| NM_001303497.1 | 6 | 5846 | NP_001290426.1 | |
| NM_001303498.1 | 5 | 5582 | NP_001290427.1 | |
| NM_001303500.1 | 4 | 5450 | NP_001290429.1 | |
| NM_152703.3 | 5 | 6222 | NP_689916.2 | |
| XM_005250193.3 | 6 | 5654 | XP_005250250.1 | |
| XM_006715890.1 | 2 | 5023 | XP_006715953.1 | |
| XM_011515903.2 | 6 | 5907 | XP_011514205.1 | |
| XM_017011822.1 | 4 | 5971 | XP_016867311.1 | |
| XM_017011823.1 | 5 | 5727 | XP_016867312.1 | |
| AB095926.1 | 5 | 6190 | ||
| AF474973.1 | 6 | 5599 | AAQ05771.1 | |
| AK026672.1 | 1 | 478 | ||
| AK097204.1 | 1 | 2404 | BAC04975.1 | |
| AK304779.1 | 1 | 3991 | ||
| AL832264.1 | 1 | 3088 | ||
| AL834232.1 | 1 | 1730 | CAD38910.1 | |
| AY195587.1 | 3 | 547 | ||
| BC127117.1 | ||||
| BC127118.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic