Assay Details
Target Gene Details
Entrez Gene ID: | 583 |
Gene Name: | Bardet-Biedl syndrome 2 |
Gene Aliases: |
BBS, RP74 |
Location: |
Chr.16:56470403-56520096 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBS2 | NM_031885.3 | NP_114091.3 | ||
| XM_005256080.2 | XP_005256137.1 | |||
| AB208905.1 | BAD92142.1 | |||
| AF342736.1 | AAK28552.1 | |||
| AK027635.1 | BAB55252.1 | |||
| AK127290.1 | 2 | 2612 | ||
| AK289604.1 | ||||
| AL834176.1 | CAD38873.2 | |||
| BC014140.2 | AAH14140.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833243 | Chr.16:56375371 - 56549461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map