Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287944 | NR_040246.1 | |||
| DA250223.1 |
Target Gene Details
Entrez Gene ID: | 55703 |
Gene Name: | RNA polymerase III subunit B |
Gene Aliases: |
C128, HLD8, INMAP, RPC2 |
Location: |
Chr.12:106357658-106510198 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3B | NM_001160708.1 | 28 | 3631 | NP_001154180.1 |
| NM_018082.5 | 28 | 3784 | NP_060552.4 | |
| AK001161.1 | 8 | 1255 | BAA91527.1 | |
| AK001250.1 | 9 | 1313 | BAA91581.1 | |
| AK090636.1 | 13 | 1856 | ||
| AK092031.1 | 18 | 2707 | ||
| AK122713.1 | 28 | 3611 | ||
| AK291635.1 | 28 | 3628 | ||
| AY092084.1 | 28 | 3615 | AAM18214.1 | |
| BC046238.1 | 28 | 3651 | AAH46238.1 | |
| BX537447.1 | 29 | 3732 | CAD97689.1 | |
| BX648712.1 | 1 | 3521 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832510 | Chr.12:106393952 - 106557362 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map