Assay Details
Target Gene Details
Entrez Gene ID: | 2110 |
Gene Name: | electron transfer flavoprotein dehydrogenase |
Gene Aliases: |
ETFQO, MADD |
Location: |
Chr.4:158672101-158708713 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETFDH | NM_001281737.1 | NP_001268666.1 | ||
| NM_001281738.1 | NP_001268667.1 | |||
| NM_004453.3 | NP_004444.2 | |||
| AK293737.1 | ||||
| AK304838.1 | ||||
| AK314803.1 | ||||
| BC011890.1 | AAH11890.1 | |||
| BX538129.1 | CAD98030.1 | |||
| EU093082.1 | ||||
| EU093083.1 | ||||
| S69232.1 | AAC60628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830125 | Chr.4:158691163 - 158848360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map