Assay Details
Target Gene Details
Entrez Gene ID: | 9411 |
Gene Name: | Rho GTPase activating protein 29 |
Gene Aliases: |
PARG1 |
Location: |
Chr.1:94168907-94275068 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP29 | NM_004815.3 | 23 | 4701 | NP_004806.3 |
| XM_006711048.3 | 22 | 4335 | XP_006711111.1 | |
| XM_011542438.2 | 23 | 4576 | XP_011540740.1 | |
| XM_011542439.2 | 23 | 4421 | XP_011540741.1 | |
| XM_011542440.2 | 22 | 4295 | XP_011540742.1 | |
| AB208873.1 | 22 | 4307 | BAD92110.1 | |
| AK310491.1 | ||||
| U90920.1 | 23 | 4515 | AAB81012.1 |
More Information
Set Membership: |
|

Genomic Map