Assay Details
Target Gene Details
Entrez Gene ID: | 3611 |
Gene Name: | integrin linked kinase |
Gene Aliases: |
HEL-S-28, ILK-1, ILK-2, P59, p59ILK |
Location: |
Chr.11:6603708-6610874 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILK | NM_001014794.2 | 3 | 351 | NP_001014794.1 |
| NM_001014795.2 | 2 | 652 | NP_001014795.1 | |
| NM_001278441.1 | 3 | 397 | NP_001265370.1 | |
| NM_001278442.1 | NP_001265371.1 | |||
| NM_004517.3 | 3 | 397 | NP_004508.1 | |
| XM_005252904.4 | 3 | 358 | XP_005252961.1 | |
| XM_005252905.2 | XP_005252962.1 | |||
| XM_011520065.1 | 3 | 357 | XP_011518367.1 | |
| XM_017017672.1 | 3 | 336 | XP_016873161.1 | |
| AB209416.1 | 1 | 4337 | BAD92653.1 | |
| AK293474.1 | ||||
| AK296628.1 | 3 | 318 | ||
| AK314843.1 | 3 | 332 | ||
| BC001554.1 | 3 | 258 | AAH01554.1 | |
| CB113885.1 | 3 | 374 | ||
| CR407673.1 | 2 | 190 | CAG28601.1 | |
| CR749220.1 | 2 | 646 | CAH18077.1 | |
| EU794625.1 | 2 | 485 | ||
| U40282.1 | 3 | 346 | AAC16892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553373 | Chr.11:6601484 - 6647097 on Build GRCh38 | Loss |
|
| nsv1035138 | Chr.11:6607812 - 6980555 on Build GRCh38 | Gain |
|
| esv996362 | Chr.11:6597567 - 6612421 on Build GRCh38 | Deletion |
|
| nsv825755 | Chr.11:6591998 - 6656613 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map