Assay Details
Target Gene Details
Entrez Gene ID: | 55861 |
Gene Name: | dysbindin domain containing 2 |
Gene Aliases: |
C20orf35, CK1BP, HSMNP1 |
Location: |
Chr.20:45405993-45410610 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBNDD2 | NM_001048221.2 | NP_001041686.1 | ||
| NM_001048222.2 | NP_001041687.1 | |||
| NM_001197139.1 | NP_001184068.1 | |||
| NM_001197140.1 | NP_001184069.1 | |||
| NM_018478.3 | 1 | 263 | NP_060948.3 | |
| AF220191.1 | 1 | 263 | AAF67656.1 | |
| BG703352.1 | ||||
| BP330715.1 | ||||
| BX335176.2 | ||||
| CB155210.1 | ||||
| CN347771.1 |
Target Gene Details
Entrez Gene ID: | 767557 |
Gene Name: | SYS1-DBNDD2 readthrough (NMD candidate) |
Gene Aliases: |
C20orf169-DBNDD2 |
Location: |
Chr.20:45363169-45410610 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYS1-DBNDD2 | NR_003189.2 | |||
| BI199175.1 | ||||
| DN994280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
| dgv4319n100 | Chr.20:45384898 - 45428463 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map