Assay Details
Target Gene Details
Entrez Gene ID: | 501 |
Gene Name: | aldehyde dehydrogenase 7 family member A1 |
Gene Aliases: |
ATQ1, EPD, PDE |
Location: |
Chr.5:126541841-126595390 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH7A1 | NM_001182.4 | NP_001173.2 | ||
| NM_001201377.1 | NP_001188306.1 | |||
| NM_001202404.1 | NP_001189333.2 | |||
| XM_011543417.2 | XP_011541719.1 | |||
| XM_017009493.1 | XP_016864982.1 | |||
| AK092507.1 | ||||
| AK295526.1 | ||||
| AK297365.1 | ||||
| AK312459.1 | ||||
| BC002515.2 | ||||
| BC071712.1 | AAH71712.1 | |||
| BC073174.1 | AAH73174.1 | |||
| S74728.1 | AAB31966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606679 | Chr.5:126424914 - 126763280 on Build GRCh38 | Gain |
|
| esv3606680 | Chr.5:126475605 - 126628219 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map