Assay Details
Target Gene Details
Entrez Gene ID: | 57619 |
Gene Name: | shroom family member 3 |
Gene Aliases: |
APXL3, MSTP013, SHRM, ShrmL |
Location: |
Chr.4:76435100-76783253 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHROOM3 | NM_020859.3 | 1 | 382 | NP_065910.3 |
| XM_011532158.2 | XP_011530460.1 | |||
| XM_011532159.1 | 1 | 369 | XP_011530461.1 | |
| AB055660.1 | 1 | 383 | BAB84689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759262 | Chr.4:76343443 - 76534252 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map