Assay Details
Target Gene Details
Entrez Gene ID: | 683 |
Gene Name: | bone marrow stromal cell antigen 1 |
Gene Aliases: |
CD157 |
Location: |
Chr.4:15686347-15774177 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BST1 | NM_004334.2 | 1 | 78 | NP_004325.2 |
| XM_005248185.1 | 1 | 78 | XP_005248242.1 | |
| XM_005248186.1 | 1 | 78 | XP_005248243.1 | |
| XM_011513878.2 | 1 | 78 | XP_011512180.1 | |
| XM_011513879.1 | 1 | 78 | XP_011512181.1 | |
| XM_011513881.2 | 1 | 1162 | XP_011512183.1 | |
| XM_017008565.1 | 1 | 78 | XP_016864054.1 | |
| XM_017008566.1 | 1 | 78 | XP_016864055.1 | |
| XM_017008567.1 | XP_016864056.1 | |||
| D21878.1 | BAA04885.1 | |||
| DA382561.1 | 1 | 79 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428439 | Chr.4:15555338 - 15747897 on Build GRCh38 | Gain+Loss |
|
| esv3599741 | Chr.4:15701308 - 15709562 on Build GRCh38 | Gain |
|
| esv2759227 | Chr.4:15442993 - 15747897 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map