Assay Details
Target Gene Details
Entrez Gene ID: | 10929 |
Gene Name: | serine and arginine rich splicing factor 8 |
Gene Aliases: |
DSM-1, SFRS2B, SRP46 |
Location: |
Chr.11:95066877-95071227 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF8 | NM_032102.3 | 1 | 2723 | NP_115285.1 |
| NR_103726.1 | 2 | 2252 | ||
| XM_017017142.1 | 2 | 2589 | XP_016872631.1 |
More Information
Set Membership: |
|

Genomic Map