Assay Details
Target Gene Details
Entrez Gene ID: | 54622 |
Gene Name: | ADP ribosylation factor like GTPase 15 |
Gene Aliases: |
ARFRP2 |
Location: |
Chr.5:53883942-54310590 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL15 | NM_019087.2 | 5 | 702 | NP_061960.1 |
| XM_011543498.2 | 7 | 807 | XP_011541800.1 | |
| XM_011543499.2 | 6 | 750 | XP_011541801.1 | |
| XM_011543500.2 | 6 | 776 | XP_011541802.1 | |
| XM_017009598.1 | 7 | 794 | XP_016865087.1 | |
| AK000058.1 | 5 | 702 | BAA90915.1 | |
| AK290886.1 | 5 | 703 | ||
| AK311482.1 | 5 | 625 | ||
| BC026093.1 | 5 | 706 | AAH26093.1 | |
| CR457225.1 | CAG33506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5666n100 | Chr.5:53231175 - 54453241 on Build GRCh38 | Gain |
|
| nsv830298 | Chr.5:53678082 - 53897568 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map