Assay Details
Target Gene Details
Entrez Gene ID: | 100996928 |
Gene Name: | C7orf55-LUC7L2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:139341132-139423457 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf55-LUC7L2 | NM_001244584.2 | NP_001231513.1 | ||
| AK297642.1 |
Target Gene Details
Entrez Gene ID: | 154791 |
Gene Name: | formation of mitochondrial complex V assembly factor 1 homolog |
Gene Aliases: |
C7orf55, HSPC268 |
Location: |
Chr.7:139340450-139346319 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FMC1 | NM_197964.4 | 2 | 550 | NP_932068.2 |
| NR_073058.1 | 3 | 423 | ||
| NR_073059.1 | 2 | 329 | ||
| NR_073060.1 | 2 | 309 | ||
| AA393224.1 | 2 | 268 | ||
| AA620996.1 | 1 | 138 | ||
| AF161386.1 | 2 | 356 | AAF28946.1 | |
| BC008467.1 | 2 | 353 | AAH08467.1 | |
| BI117784.1 | 2 | 550 | ||
| BM672187.1 | 1 | 153 | ||
| BU852633.1 | 2 | 333 | ||
| CB125801.1 |
Target Gene Details
Entrez Gene ID: | 51631 |
Gene Name: | LUC7 like 2, pre-mRNA splicing factor |
Gene Aliases: |
CGI-59, CGI-74, LUC7B2 |
Location: |
Chr.7:139340359-139423457 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUC7L2 | NM_001244585.1 | NP_001231514.1 | ||
| AK298166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map