Assay Details
Target Gene Details
Entrez Gene ID: | 6336 |
Gene Name: | sodium voltage-gated channel alpha subunit 10 |
Gene Aliases: |
FEPS2, Nav1.8, PN3, SNS, hPN3 |
Location: |
Chr.3:38697110-38794010 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN10A | NM_001293306.2 | 26 | 4553 | NP_001280235.2 |
| NM_001293307.2 | 25 | 4262 | NP_001280236.2 | |
| NM_006514.3 | 26 | 4556 | NP_006505.3 | |
| XM_005265371.3 | 26 | 4565 | XP_005265428.1 | |
| XM_011533993.2 | 26 | 4562 | XP_011532295.1 | |
| XM_011533994.2 | 25 | 4271 | XP_011532296.1 | |
| AF117907.1 | 26 | 4556 | AAD30863.1 |
More Information
Set Membership: |
|

Genomic Map