Assay Details
Target Gene Details
Entrez Gene ID: | 10929 |
Gene Name: | serine and arginine rich splicing factor 8 |
Gene Aliases: |
DSM-1, SFRS2B, SRP46 |
Location: |
Chr.11:95066877-95071227 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF8 | NM_032102.3 | 1 | 976 | NP_115285.1 |
| NR_103726.1 | 1 | 976 | ||
| XM_017017142.1 | 1 | 938 | XP_016872631.1 | |
| AF031166.1 | 1 | 596 | AAK54351.1 | |
| AK023379.1 | 1 | 862 | ||
| AK309776.1 | ||||
| AK312514.1 | 1 | 865 | ||
| BC057783.1 | 2 | 666 | AAH57783.1 | |
| CV805885.1 | 1 | 359 |
More Information
Set Membership: |
|

Genomic Map