Assay Details
Target Gene Details
Entrez Gene ID: | 23481 |
Gene Name: | pescadillo ribosomal biogenesis factor 1 |
Gene Aliases: |
PES |
Location: |
Chr.22:30576625-30607013 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PES1 | NM_001243225.1 | 15 | 1988 | NP_001230154.1 |
| NM_001282327.1 | 17 | 2401 | NP_001269256.1 | |
| NM_001282328.1 | 17 | 2448 | NP_001269257.1 | |
| NM_014303.3 | 15 | 2003 | NP_055118.1 | |
| XM_006724173.1 | 15 | 1956 | XP_006724236.1 | |
| XM_017028678.1 | 12 | 3821 | XP_016884167.1 | |
| AK096320.1 | 17 | 2420 | ||
| AK125778.1 | 2 | 1451 | BAC86285.1 | |
| AK127185.1 | 17 | 2402 | ||
| BC032489.1 | 15 | 1960 | AAH32489.1 | |
| BQ576244.1 | 1 | 310 | ||
| CR456539.1 | 15 | 1906 | CAG30425.1 | |
| U78310.1 | 15 | 1954 | AAB61140.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588887 | Chr.22:30557534 - 30599369 on Build GRCh38 | Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map