Assay Details
Target Gene Details
Entrez Gene ID: | 196951 |
Gene Name: | family with sequence similarity 227 member B |
Gene Aliases: |
C15orf33 |
Location: |
Chr.15:49326997-49620931 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM227B | NM_152647.2 | 13 | 1459 | NP_689860.2 |
| XM_005254213.3 | 13 | 1536 | XP_005254270.1 | |
| XM_005254214.3 | 13 | 1469 | XP_005254271.1 | |
| XM_005254215.3 | 12 | 1500 | XP_005254272.1 | |
| XM_005254216.3 | 12 | 1434 | XP_005254273.1 | |
| XM_005254219.3 | XP_005254276.1 | |||
| XM_006720423.2 | 14 | 1501 | XP_006720486.1 | |
| XM_006720424.2 | 13 | 1536 | XP_006720487.1 | |
| XM_006720426.2 | 11 | 1395 | XP_006720489.1 | |
| XM_006720427.3 | 14 | 1507 | XP_006720490.1 | |
| XM_011521319.2 | 13 | 3419 | XP_011519621.1 | |
| XM_011521320.1 | 11 | 911 | XP_011519622.1 | |
| XM_011521321.2 | XP_011519623.1 | |||
| XM_011521322.1 | 10 | 875 | XP_011519624.1 | |
| XM_017021990.1 | 10 | 1293 | XP_016877479.1 | |
| XM_017021991.1 | 14 | 1576 | XP_016877480.1 | |
| XM_017021992.1 | 11 | 1224 | XP_016877481.1 | |
| XM_017021993.1 | 10 | 1293 | XP_016877482.1 | |
| XM_017021994.1 | 9 | 1194 | XP_016877483.1 | |
| XM_017021995.1 | XP_016877484.1 | |||
| XM_017021996.1 | 12 | 984 | XP_016877485.1 | |
| AK023699.1 | ||||
| AK057362.1 | 13 | 1457 | BAB71451.1 |
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALK2 | XM_005254279.3 | 11 | 1508 | XP_005254336.1 |
| XM_005254280.3 | 11 | 1599 | XP_005254337.1 | |
| XM_005254284.3 | 10 | 1500 | XP_005254341.1 | |
| XM_006720462.3 | 11 | 1489 | XP_006720525.1 | |
| XM_006720463.3 | 10 | 1389 | XP_006720526.1 | |
| XM_006720464.3 | 9 | 1236 | XP_006720527.1 | |
| XM_011521441.2 | 12 | 1712 | XP_011519743.1 | |
| XM_017022063.1 | 9 | 1347 | XP_016877552.1 | |
| XM_017022064.1 | 9 | 1565 | XP_016877553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636427 | Chr.15:49359900 - 49397828 on Build GRCh38 | Loss |
|
| nsv833002 | Chr.15:49329203 - 49489602 on Build GRCh38 | Gain |
|
| nsv442375 | Chr.15:49360327 - 49391885 on Build GRCh38 | Loss |
|
| esv3636426 | Chr.15:49354085 - 49408851 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map