Assay Details
Target Gene Details
Entrez Gene ID: | 63915 |
Gene Name: | biogenesis of lysosomal organelles complex 1 subunit 5 |
Gene Aliases: |
BLOS5, MU, MUTED |
Location: |
Chr.6:8013567-8064414 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLOC1S5 | NM_001199322.1 | 6 | 753 | NP_001186251.1 |
| NM_001199323.1 | 4 | 520 | NP_001186252.1 | |
| NM_201280.2 | 5 | 650 | NP_958437.1 | |
| AF426434.1 | 5 | 650 | AAL99385.1 | |
| AK025442.1 | 5 | 603 | ||
| AK301142.1 | 5 | 651 | ||
| AK311385.1 | 2 | 501 | ||
| BC032438.1 | 5 | 613 | AAH32438.1 | |
| BC119644.1 | 5 | 635 | ||
| BC119645.1 | 6 | 735 | ||
| BM829259.1 | 4 | 491 | ||
| BX647336.1 | 6 | 822 | ||
| CR749569.1 | 5 | 625 | CAH18364.1 |
Target Gene Details
Entrez Gene ID: | 100526836 |
Gene Name: | BLOC1S5-TXNDC5 readthrough (NMD candidate) |
Gene Aliases: |
MUTED-TXNDC5 |
Location: |
Chr.6:7881250-8064414 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLOC1S5-TXNDC5 | NR_037616.1 | |||
| BC052310.1 | AAH52310.1 |
Target Gene Details
Entrez Gene ID: | 100526837 |
Gene Name: | EEF1E1-BLOC1S5 readthrough (NMD candidate) |
Gene Aliases: |
EEF1E1-MUTED |
Location: |
Chr.6:8013567-8102595 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF1E1-BLOC1S5 | NR_037618.1 | 7 | 958 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600924 | Chr.6:7823346 - 8206902 on Build GRCh38 | Gain |
|
| esv2731544 | Chr.6:7817430 - 8348230 on Build GRCh38 | Deletion |
|
| esv3576096 | Chr.6:7993744 - 8022398 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map