Assay Details
Target Gene Details
Entrez Gene ID: | 1368 |
Gene Name: | carboxypeptidase M |
Gene Aliases: |
- |
Location: |
Chr.12:68842200-68963240 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPM | NM_001005502.2 | 9 | 2332 | NP_001005502.1 |
| NM_001874.4 | 9 | 2364 | NP_001865.1 | |
| NM_198320.3 | 9 | 2350 | NP_938079.1 | |
| AF368463.1 | 9 | 2322 | AAK69717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629859 | Chr.12:68854895 - 68855816 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map