Assay Details
Target Gene Details
Entrez Gene ID: | 8405 |
Gene Name: | speckle type BTB/POZ protein |
Gene Aliases: |
BTBD32, TEF2 |
Location: |
Chr.17:49598884-49678163 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPOP | NM_001007226.1 | NP_001007227.1 | ||
| NM_001007227.1 | NP_001007228.1 | |||
| NM_001007228.1 | NP_001007229.1 | |||
| NM_001007229.1 | NP_001007230.1 | |||
| NM_001007230.1 | NP_001007231.1 | |||
| NM_003563.3 | NP_003554.1 | |||
| XM_005257723.4 | XP_005257780.1 | |||
| XM_005257724.4 | XP_005257781.1 | |||
| XM_017025204.1 | XP_016880693.1 | |||
| AJ000644.1 | CAA04199.1 | |||
| AK222589.1 | BAD96309.1 | |||
| AK293045.1 | ||||
| AK293587.1 | ||||
| AK300616.1 | ||||
| AK303268.1 | ||||
| AK312691.1 | ||||
| AK316366.1 | ||||
| BC001269.1 | AAH01269.1 | |||
| BC003385.1 | AAH03385.1 |
More Information
Set Membership: |
|

Genomic Map