Assay Details
Target Gene Details
| Entrez Gene ID: | 51652 | 
| Gene Name: | charged multivesicular body protein 3 | 
| Gene Aliases: | CGI-149, NEDF, VPS24 | 
| Location: | Chr.2:86503430-86563497 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CHMP3 | NM_001005753.2 | 5 | 886 | NP_001005753.1 | 
| NM_001193517.1 | 6 | 827 | NP_001180446.1 | |
| NM_016079.3 | 6 | 947 | NP_057163.1 | |
| NR_036454.1 | 4 | 706 | ||
| AF151907.1 | 6 | 816 | AAD34144.1 | |
| AK025260.1 | 6 | 845 | ||
| AK129502.1 | 6 | 704 | ||
| AK290725.1 | 5 | 777 | ||
| AK315835.1 | 5 | 830 | ||
| AY364249.1 | AAQ76808.1 | |||
| BC004419.1 | 6 | 845 | AAH04419.1 | 
Target Gene Details
| Entrez Gene ID: | 100526767 | 
| Gene Name: | RNF103-CHMP3 readthrough | 
| Gene Aliases: | RNF103-VPS24 | 
| Location: | Chr.2:86503430-86721122 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 8 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| RNF103-CHMP3 | NM_001198954.1 | 8 | 1128 | NP_001185883.1 | 
| AK294389.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv2813 | Chr.2:86480671 - 86514180 on Build GRCh38 | Insertion |  CHMP3  KDM3A  RNF103-CHMP3 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top