Assay Details
Target Gene Details
Entrez Gene ID: | 2553 |
Gene Name: | GA binding protein transcription factor beta subunit 1 |
Gene Aliases: |
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2 |
Location: |
Chr.15:50277192-50355429 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABPB1 | NM_001320910.1 | 10 | 2928 | NP_001307839.1 |
| NM_001320915.1 | 10 | 2925 | NP_001307844.1 | |
| NM_005254.5 | 9 | 2883 | NP_005245.2 | |
| NM_016654.4 | 9 | 2847 | NP_057738.1 | |
| XM_005254274.3 | 9 | 2591 | XP_005254331.1 | |
| XM_011521426.2 | 9 | 2844 | XP_011519728.1 | |
| XM_017022054.1 | 10 | 2584 | XP_016877543.1 | |
| AI631183.1 | 1 | 187 | ||
| BC036080.1 | 9 | 2641 | AAH36080.1 | |
| BE644609.1 | 1 | 117 | ||
| U13045.1 | 9 | 2628 | AAA65707.1 | |
| U13046.1 | 9 | 2592 | AAA65708.1 |
More Information
Set Membership: |
|

Genomic Map