Assay Details
Target Gene Details
Entrez Gene ID: | 119391 |
Gene Name: | glutathione S-transferase omega 2 |
Gene Aliases: |
GSTO 2-2, bA127L20.1 |
Location: |
Chr.10:104267626-104304948 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTO2 | NM_001191013.1 | NP_001177942.1 | ||
| NM_001191014.1 | NP_001177943.1 | |||
| NM_001191015.1 | NP_001177944.1 | |||
| NM_183239.1 | NP_899062.1 | |||
| XM_006717624.3 | XP_006717687.1 | |||
| XM_011539270.2 | XP_011537572.1 | |||
| XM_011539271.1 | XP_011537573.1 | |||
| XM_011539272.2 | XP_011537574.1 | |||
| XM_011539273.2 | XP_011537575.1 | |||
| XM_017015670.1 | XP_016871159.1 | |||
| XM_017015671.1 | XP_016871160.1 | |||
| AK291886.1 | ||||
| AK296266.1 | ||||
| AK297522.1 | ||||
| AK300508.1 | ||||
| AY191318.1 | AAO23573.1 | |||
| AY209189.1 | AAP47743.1 | |||
| AY350731.1 | AAR02452.1 | |||
| BC056918.1 | AAH56918.1 | |||
| BM708208.1 | ||||
| DC341647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831971 | Chr.10:104120100 - 104301309 on Build GRCh38 | Gain |
|
| nsv552062 | Chr.10:104254126 - 104282744 on Build GRCh38 | Loss |
|
| nsv1039166 | Chr.10:104260183 - 104302423 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map