Assay Details
Target Gene Details
Entrez Gene ID: | 27161 |
Gene Name: | argonaute 2, RISC catalytic component |
Gene Aliases: |
EIF2C2, Q10 |
Location: |
Chr.8:140520158-140642406 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO2 | NM_001164623.1 | 18 | 2941 | NP_001158095.1 |
| NM_012154.3 | 19 | 3043 | NP_036286.2 | |
| XM_011516965.2 | 19 | 6119 | XP_011515267.1 | |
| XM_011516966.2 | 19 | 3860 | XP_011515268.1 | |
| XM_011516968.2 | 19 | 3194 | XP_011515270.2 | |
| XM_017013317.1 | 18 | 2812 | XP_016868806.1 | |
| AF121255.2 | 8 | 1556 | AAF13034.2 | |
| BC018727.1 | 14 | 2290 | AAH18727.2 | |
| BC125213.1 | 18 | 2849 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2737756 | Chr.8:140419230 - 141917755 on Build GRCh38 | Deletion |
|
| nsv1016282 | Chr.8:140453461 - 140537127 on Build GRCh38 | Gain |
|
| nsv1126125 | Chr.8:140419236 - 141917668 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map