Assay Details
Target Gene Details
Entrez Gene ID: | 27347 |
Gene Name: | serine/threonine kinase 39 |
Gene Aliases: |
DCHT, PASK, SPAK |
Location: |
Chr.2:167954020-168247595 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK39 | NM_013233.2 | NP_037365.2 | ||
| XM_005246465.2 | XP_005246522.1 | |||
| AF099989.1 | AAC72238.1 | |||
| AK126343.1 | ||||
| BF516344.1 | ||||
| BX647113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583596 | Chr.2:167764495 - 167960492 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map