Assay Details
Target Gene Details
Entrez Gene ID: | 5305 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase type 2 alpha |
Gene Aliases: |
PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK |
Location: |
Chr.10:22534837-22714574 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP4K2A | NM_005028.4 | NP_005019.2 | ||
| XM_006717450.2 | XP_006717513.1 | |||
| XM_011519496.1 | XP_011517798.1 | |||
| XM_017016330.1 | XP_016871819.1 | |||
| XM_017016331.1 | XP_016871820.1 | |||
| XM_017016332.1 | XP_016871821.1 | |||
| AK294817.1 | ||||
| AK294867.1 | ||||
| BC018034.1 | AAH18034.1 | |||
| S78798.1 | AAB35041.1 | |||
| U14957.2 | AAA64835.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs112020551] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map