Assay Details
Target Gene Details
Entrez Gene ID: | 55922 |
Gene Name: | NFKB repressing factor |
Gene Aliases: |
ITBA4, NRF |
Location: |
Chr.X:119588337-119606464 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKRF | NM_001173487.1 | 4 | 951 | NP_001166958.1 |
| NM_001173488.1 | 2 | 1337 | NP_001166959.1 | |
| NM_017544.3 | 3 | 853 | NP_060014.2 | |
| XM_011531365.2 | 4 | 1483 | XP_011529667.1 | |
| AJ011812.2 | 2 | 1337 | CAB56459.1 | |
| AL539002.3 | 4 | 880 | ||
| AY208891.1 | 3 | 853 | AAP43025.1 | |
| BC040379.1 | 3 | 835 | AAH40379.1 | |
| BC047878.2 | 3 | 820 | AAH47878.1 | |
| BC068514.1 | 3 | 831 | AAH68514.1 | |
| EF165092.1 | 1 | 411 | ||
| EF165094.1 | ||||
| EF216688.1 | 2 | 1337 | ||
| EF216689.1 | 2 | 1337 |
More Information
Set Membership: |
|

Genomic Map