Assay Details
Target Gene Details
Entrez Gene ID: | 11343 |
Gene Name: | monoglyceride lipase |
Gene Aliases: |
HU-K5, HUK5, MAGL, MGL |
Location: |
Chr.3:127689062-127823250 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGLL | NM_001003794.2 | 8 | 1489 | NP_001003794.1 |
| NM_001256585.1 | 7 | 1806 | NP_001243514.1 | |
| NM_007283.6 | 8 | 1896 | NP_009214.1 | |
| XM_011512377.1 | 9 | 1946 | XP_011510679.1 | |
| XM_011512378.1 | 9 | 1567 | XP_011510680.1 | |
| XM_011512379.2 | 10 | 1683 | XP_011510681.1 | |
| XM_011512382.1 | 8 | 1509 | XP_011510684.1 | |
| XM_011512383.2 | 8 | 1470 | XP_011510685.1 | |
| XM_017005664.1 | 9 | 2057 | XP_016861153.1 | |
| XM_017005665.1 | 7 | 1619 | XP_016861154.1 | |
| XM_017005666.1 | 7 | 1399 | XP_016861155.1 | |
| AK093841.1 | 1 | 237 | ||
| AL833254.1 | 1 | 272 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460848 | Chr.3:127667659 - 127702011 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map