Assay Details
Target Gene Details
Entrez Gene ID: | 10239 |
Gene Name: | adaptor related protein complex 3 sigma 2 subunit |
Gene Aliases: |
AP3S3, sigma3b |
Location: |
Chr.15:89830599-89894385 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3S2 | NM_005829.4 | 6 | 1439 | NP_005820.1 |
| NR_023361.1 | 7 | 1558 | ||
| NR_037582.1 | 6 | 1435 | ||
| AK294712.1 | 7 | 1150 | ||
| BC002785.2 | 6 | 1024 | AAH02785.1 | |
| BC007773.2 | 6 | 1023 | AAH07773.1 | |
| BC010020.2 | 6 | 1051 | AAH10020.1 | |
| BX098014.1 | 1 | 115 | ||
| X99459.1 | 6 | 1033 | CAA67824.1 |
Target Gene Details
Entrez Gene ID: | 100526783 |
Gene Name: | C15orf38-AP3S2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:89830599-89912990 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf38-AP3S2 | NM_001199058.1 | 10 | 1761 | NP_001185987.1 |
| AK000495.1 | 10 | 1653 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1661 | Chr.15:89804782 - 89839712 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map