Assay Details
Target Gene Details
Entrez Gene ID: | 84656 |
Gene Name: | glyoxylate reductase 1 homolog |
Gene Aliases: |
BM045, HIBDL, N-PAC, NP60 |
Location: |
Chr.16:4803203-4847388 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLYR1 | NM_001308096.1 | 16 | 2403 | NP_001295025.1 |
| NM_001324096.1 | NP_001311025.1 | |||
| NM_001324097.1 | NP_001311026.1 | |||
| NM_001324098.1 | 15 | 2456 | NP_001311027.1 | |
| NM_032569.3 | 16 | 2421 | NP_115958.2 | |
| NR_136695.1 | ||||
| NR_136696.1 | ||||
| NR_136697.1 | ||||
| NR_136698.1 | ||||
| NR_136699.1 | ||||
| NR_136700.1 | 15 | 2573 | ||
| XM_005255639.4 | 14 | 2171 | XP_005255696.2 | |
| XM_011522716.2 | 16 | 2549 | XP_011521018.1 | |
| XM_011522717.1 | 17 | 2606 | XP_011521019.1 | |
| XM_017023786.1 | 16 | 2499 | XP_016879275.1 | |
| XM_017023787.1 | 15 | 2433 | XP_016879276.1 | |
| XM_017023788.1 | 17 | 2588 | XP_016879277.1 | |
| XM_017023789.1 | 16 | 2555 | XP_016879278.1 | |
| AF217521.1 | 1 | 273 | AAF67632.1 | |
| AF244907.1 | 16 | 2402 | AAQ14242.1 | |
| BC032855.1 | 16 | 2412 | AAH32855.1 | |
| BC047223.1 | 16 | 2401 | AAH47223.1 | |
| BC064940.1 | AAH64940.1 | |||
| CR933687.1 | 3 | 704 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26756 | Chr.16:4772529 - 4855231 on Build GRCh38 | Gain+Loss |
|
| esv2422516 | Chr.16:4775920 - 5160979 on Build GRCh38 | Deletion |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map