Assay Details
Target Gene Details
Entrez Gene ID: | 54617 |
Gene Name: | INO80 complex subunit |
Gene Aliases: |
INO80A, INOC1, hINO80 |
Location: |
Chr.15:40978880-41117081 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80 | NM_017553.2 | 36 | 6170 | NP_060023.1 |
| NR_104038.1 | 35 | 6042 | ||
| XM_011521686.2 | 22 | 3981 | XP_011519988.1 | |
| AB033085.1 | 37 | 5831 | ||
| AK002176.1 | 8 | 2334 | BAA92122.1 | |
| AK024777.1 | 1 | 976 | BAB14997.1 | |
| AL137280.1 | 5 | 1950 | CAB70675.1 | |
| BC146785.1 | 37 | 5843 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053462 | Chr.15:40977869 - 41103896 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map