Assay Details
Target Gene Details
Entrez Gene ID: | 121506 |
Gene Name: | endoplasmic reticulum protein 27 |
Gene Aliases: |
C12orf46, PDIA8 |
Location: |
Chr.12:14914027-14938549 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP27 | NM_001300784.1 | 5 | 962 | NP_001287713.1 |
| NM_152321.3 | 7 | 1155 | NP_689534.1 | |
| AA772096.1 | 1 | 416 | ||
| AK056677.1 | 7 | 1155 | BAB71251.1 | |
| AY358536.1 | 7 | 1135 | AAQ88900.1 | |
| BC030218.1 | 7 | 1127 | AAH30218.1 | |
| BE501761.1 | 1 | 426 | ||
| BQ574479.1 | 1 | 417 | ||
| CD389092.1 | 5 | 837 |
More Information
Set Membership: |
|

Genomic Map