Assay Details
Target Gene Details
Entrez Gene ID: | 64232 |
Gene Name: | membrane spanning 4-domains A5 |
Gene Aliases: |
CD20-L2, CD20L2, TETM4 |
Location: |
Chr.11:60429589-60447792 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A5 | NM_023945.2 | NP_076434.2 | ||
| AB013103.1 | BAB18739.1 | |||
| AF237907.1 | AAK37416.1 | |||
| AF321127.1 | AAK01641.1 | |||
| BC029884.1 | AAH29884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744569 | Chr.11:60310239 - 60485227 on Build GRCh38 | Deletion |
|
| nsv508635 | Chr.11:60422385 - 60487633 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map