Assay Details
Target Gene Details
Entrez Gene ID: | 85438 |
Gene Name: | calcium binding protein, spermatid associated 1 |
Gene Aliases: |
C4orf35, CLPH, NYD-SP26 |
Location: |
Chr.4:70334954-70337116 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABS1 | NM_033122.3 | 1 | 258 | NP_149113.3 |
| AF380838.1 | 1 | 228 | AAK57455.1 | |
| AK057459.1 | 1 | 231 | BAB71497.1 | |
| AK315022.1 | 1 | 231 | ||
| BC046111.1 | 1 | 220 | AAH46111.1 | |
| DB027445.1 | 1 | 269 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
| nsv1006332 | Chr.4:70295888 - 70412931 on Build GRCh38 | Loss |
|
| nsv829964 | Chr.4:70274771 - 70439225 on Build GRCh38 | Gain |
|
| nsv461554 | Chr.4:70296786 - 70399455 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map