Assay Details
Target Gene Details
Entrez Gene ID: | 84530 |
Gene Name: | serine/arginine repetitive matrix 4 |
Gene Aliases: |
KIAA1853, MU-MB-2.76, nSR100 |
Location: |
Chr.12:118981495-119163051 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRM4 | NM_194286.3 | 1 | 213 | NP_919262.2 |
| AB058756.1 | 1 | 117 | ||
| AK291361.1 | 1 | 213 | ||
| BC152471.1 | 1 | 128 | ||
| BC167162.1 | 1 | 117 | ||
| BC167163.1 | 1 | 117 |
More Information
Set Membership: |
|

Genomic Map