Assay Details
Target Gene Details
Entrez Gene ID: | 341947 |
Gene Name: | cytochrome c oxidase subunit 8C |
Gene Aliases: |
COX8-3 |
Location: |
Chr.14:93347191-93348354 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX8C | NM_182971.2 | 1 | 39 | NP_892016.1 |
| AY161004.1 | 1 | 39 | AAO26193.1 | |
| BC101125.2 | AAI01126.1 | |||
| BC101126.2 | AAI01127.1 |
Target Gene Details
Entrez Gene ID: | 57578 |
Gene Name: | unc-79 homolog (C. elegans) |
Gene Aliases: |
KIAA1409 |
Location: |
Chr.14:93333219-93710473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC79 | NM_020818.3 | NP_065869.3 | ||
| XM_011537027.2 | XP_011535329.1 | |||
| XM_011537028.2 | XP_011535330.1 | |||
| AK125327.1 | BAC86131.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map