Assay Details
Target Gene Details
Entrez Gene ID: | 2800 |
Gene Name: | golgin A1 |
Gene Aliases: |
golgin-97 |
Location: |
Chr.9:124878294-124941114 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA1 | NM_002077.3 | 23 | 4630 | NP_002068.1 |
| XM_005251929.3 | 23 | 4562 | XP_005251986.1 | |
| XM_006717062.3 | 23 | 4626 | XP_006717125.1 | |
| XM_006717063.3 | 23 | 4597 | XP_006717126.1 | |
| AB208858.1 | 18 | 3983 | BAD92095.1 | |
| AW237629.1 | 1 | 205 | ||
| BC032853.1 | 23 | 4600 | AAH32853.1 | |
| U51587.1 | 23 | 4622 | AAB81549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573384 | Chr.9:124877849 - 124881830 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map