Assay Details
Target Gene Details
Entrez Gene ID: | 9925 |
Gene Name: | zinc finger and BTB domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.9:37438102-37465410 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB5 | NM_014872.2 | 2 | 2733 | NP_055687.1 |
| XM_005251634.1 | 2 | 3577 | XP_005251691.1 | |
| AB002352.1 | 2 | 2722 |
More Information
Set Membership: |
|

Genomic Map