Assay Details
Target Gene Details
Entrez Gene ID: | 4193 |
Gene Name: | MDM2 proto-oncogene |
Gene Aliases: |
ACTFS, HDMX, hdm2 |
Location: |
Chr.12:68808149-68845544 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDM2 | NM_001145339.2 | 1 | 143 | NP_001138811.1 |
| NM_002392.5 | 1 | 143 | NP_002383.2 | |
| XM_006719400.3 | 1 | 166 | XP_006719463.1 | |
| AK290341.1 | 1 | 93 | ||
| BE296905.1 | 1 | 108 | ||
| HY174841.1 | 1 | 145 | ||
| M92424.1 | 1 | 123 | AAA60568.1 | |
| Z12020.1 | 1 | 123 | CAA78055.1 |
More Information
Set Membership: |
|

Genomic Map