Assay Details
Target Gene Details
Entrez Gene ID: | 145781 |
Gene Name: | GRINL1A complex locus 1 |
Gene Aliases: |
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom |
Location: |
Chr.15:57591904-57717557 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCOM1 | NM_001018090.6 | NP_001018100.1 | ||
| NM_001018091.6 | NP_001018101.1 | |||
| NM_001285900.3 | NP_001272829.1 | |||
| NR_104367.2 | ||||
| NR_104368.2 | ||||
| NR_104369.2 | ||||
| NR_104370.2 | ||||
| NR_104371.3 | ||||
| AY207007.1 | AAO39707.1 | |||
| AY208913.1 | AAP41549.1 | |||
| AY331564.1 | AAQ76825.1 | |||
| AY333779.1 | AAQ76826.1 | |||
| AY334560.1 | AAQ76827.1 | |||
| AY334561.1 | AAQ76828.1 | |||
| AY350748.1 | AAQ82541.1 | |||
| AY353058.1 | AAQ76834.1 | |||
| JF419331.1 |
Target Gene Details
Entrez Gene ID: | 81488 |
Gene Name: | RNA polymerase II subunit M |
Gene Aliases: |
GCOM1, GRINL1A, Gdown, Gdown1 |
Location: |
Chr.15:57706521-57717557 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2M | NM_001018102.2 | NP_001018112.1 | ||
| NM_015532.4 | NP_056347.1 | |||
| AF326773.1 | AAK92284.2 | |||
| AK074767.1 | BAC11193.1 | |||
| AK074955.1 | BAC11313.1 | |||
| AK128618.1 | BAC87533.1 | |||
| AK223571.1 | BAD97291.1 | |||
| AK308873.1 | ||||
| AL050091.1 | CAB43263.1 | |||
| AY339380.1 | AAQ82540.1 | |||
| AY353061.1 | AAQ76837.1 | |||
| BC001510.2 | AAH01510.1 | |||
| BQ072611.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045611 | Chr.15:57058767 - 57778609 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map