Assay Details
Target Gene Details
Entrez Gene ID: | 8807 |
Gene Name: | interleukin 18 receptor accessory protein |
Gene Aliases: |
ACPL, CD218b, CDw218b, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1R-7, IL-1R7, IL-1RAcPL, IL18RB |
Location: |
Chr.2:102418551-102452568 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL18RAP | NM_003853.3 | 4 | 671 | NP_003844.1 |
| XM_011512087.2 | XP_011510389.1 | |||
| AF077346.1 | 4 | 666 | AAC72196.1 | |
| AK300026.1 | 4 | 771 | ||
| BC069630.1 | 2 | 268 | AAH69630.1 | |
| BC106764.2 | AAI06765.1 | |||
| BC106765.2 | 3 | 395 | AAI06766.1 | |
| BC137474.1 | 2 | 211 | ||
| BC137475.1 | 2 | 211 | ||
| DQ116957.1 | 2 | 183 | AAZ52551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953499 | Chr.2:102418141 - 102431540 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map